accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOH3CNHF(E)

CAS RN 18529-17-4

IUPAC Name: ethyl (2E)-3-[(4-fluorophenyl)amino]but-2-enoate
Synonyms: ethyl (2E)-3-[(4-fluorophenyl)amino]but-2-enoate
Molecular Weight (g/mol): 223.25
Molecular Formula: C12H14FNO2
InChi Key: BUDOLSNUNUYBLF-CMDGGOBGSA-N
SMILES: CCOC(=O)\C=C(/C)NC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results