accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1850-14-2

CH3OOH3COH3COH3C

CAS RN 1850-14-2

IUPAC Name: tetramethoxymethane
Synonyms: tetramethoxymethane
Molecular Weight (g/mol): 136.15
Molecular Formula: C5H12O4
InChi Key: AHJWSRRHTXRLAQ-UHFFFAOYSA-N
SMILES: COC(OC)(OC)OC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)