accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHNS

CAS RN 184847-97-0

IUPAC Name: [3-(1,3-thiazol-2-yl)phenyl]methanol
Synonyms: [3-(1,3-thiazol-2-yl)phenyl]methanol
Molecular Weight (g/mol): 191.25
Molecular Formula: C10H9NOS
InChi Key: VDFDKGHCZKGOLZ-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results