accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 184000-11-1

HOBOHOO

CAS RN 184000-11-1

IUPAC Name: {4-[(benzyloxy)carbonyl]phenyl}boronic acid
Synonyms: 4-[(benzyloxy)carbonyl]phenylboronic acid
Molecular Weight (g/mol): 256.06
Molecular Formula: C14H13BO4
InChi Key: UXSDJFGNRVVDFM-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results