CAS RN 1839099-22-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-N-Desacryloyl N-Propionyl Ibrutinib, TRC
CAS: 1839099-22-7 Molecular Formula: C25 H26 N6 O2 Molecular Weight (g/mol): 442.51 Synonym: 1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-1-propanone IUPAC Name: 1-[(3R)-3-[4-amino-3-(4-phenoxyphenyl)pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidyl]propan-1-one SMILES: CCC(=O)N1CCC[C@H](C1)n2nc(c3ccc(Oc4ccccc4)cc3)c5c(N)ncnc25