accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COH2C

CAS RN 1838-94-4

IUPAC Name: 2-ethenyl-2-methyloxirane
Synonyms: 2-ethenyl-2-methyloxirane
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: FVCDMHWSPLRYAB-UHFFFAOYNA-N
SMILES: CC1(CO1)C=C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)