accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNOCH3

CAS RN 18370-11-1

IUPAC Name: N,4-dimethylbenzamide
Synonyms: N,4-dimethylbenzamide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: FZIOOTTWDRFBKU-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results