accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1837-55-4

NNClCl

CAS RN 1837-55-4

IUPAC Name: 3,5-dichloropyridazine
Synonyms: 3,5-dichloropyridazine
Molecular Weight (g/mol): 148.97
Molecular Formula: C4H2Cl2N2
InChi Key: JZSAUQMXKHBZEO-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=NN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results