CAS RN 1835705-57-1
CAS RN 1835705-57-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress (S,R,S)-AHPC-PEG2-C4-Cl
MedChemExpress (S,R,S)-AHPC-PEG2-C4-Cl (VH032-PEG2-C4-Cl) is a conjugate of ligands for E3 and 13-atom-length linker.