accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1835-11-6

H3COOCH3O

CAS RN 1835-11-6

IUPAC Name: 1-[4-(benzyloxy)-3-methoxyphenyl]ethan-1-one
Synonyms: 1-[4-(benzyloxy)-3-methoxyphenyl]ethanone
Molecular Weight (g/mol): 256.3
Molecular Formula: C16H16O3
InChi Key: HRUAWSQBQLYDKH-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1OCC1=CC=CC=C1)C(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)