accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OHO

CAS RN 183210-33-5

IUPAC Name: (5-methyl-2-phenylfuran-3-yl)methanol
Synonyms: (5-methyl-2-phenylfuran-3-yl)methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: VZFFMWAZPODJRX-UHFFFAOYSA-N
SMILES: CC1=CC(CO)=C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results