accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1829-28-3

H3COOI

CAS RN 1829-28-3

IUPAC Name: ethyl 2-iodobenzoate
Synonyms: ethyl 2-iodobenzoate
Molecular Weight (g/mol): 276.07
Molecular Formula: C9H9IO2
InChi Key: QOUFDDUDXYJWHV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1I
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results