accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CO(R)O(R)OH(R)OH(S)HO(R)HO

CAS RN 1824-94-8

IUPAC Name: (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Synonyms: methyl β-galactoside methyl β-D-galactoside 3-O-methyl-D-galactose GalβOMe methyl galactoside 3-o-methyl-d-galactose methylgalactoside
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H14O6
InChi Key: HOVAGTYPODGVJG-VOQCIKJUSA-N
SMILES: CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)