accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 182247-45-6

H3COO(S)OO

CAS RN 182247-45-6

IUPAC Name: (3S)-3-benzyl-4-methoxy-4-oxobutanoate
Synonyms: (3S)-3-benzyl-4-methoxy-4-oxobutanoate
Molecular Weight (g/mol): 221.23
Molecular Formula: C12H13O4
InChi Key: BUNMUVFKMIOEQU-JTQLQIEISA-M
SMILES: COC(=O)[C@H](CC([O-])=O)CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results