accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNNH2OH2N

CAS RN 18213-75-7

IUPAC Name: 5-amino-1-methyl-1H-pyrazole-4-carboxamide
Synonyms: 5-amino-1-methylpyrazole-4-carboxamide
Molecular Weight (g/mol): 140.15
Molecular Formula: C5H8N4O
InChi Key: JGSQVTVXGXOSCH-UHFFFAOYSA-N
SMILES: CN1N=CC(C(N)=O)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results