CAS RN 18166-37-5
IUPAC Name:
triethoxy(hexyl)silane
Synonyms:
triethoxy(hexyl)silane
Molecular Weight (g/mol):
248.44
Molecular Formula:
C12H28O3Si
InChi Key:
WUMSTCDLAYQDNO-UHFFFAOYSA-N
SMILES:
CCCCCC[Si](OCC)(OCC)OCC
1
–
1
of
1
results
1
n-Hexyltriethoxysilane, 97%
CAS: 18166-37-5 Molecular Formula: C12H28O3Si Molecular Weight (g/mol): 248.44 MDL Number: MFCD00087744 InChI Key: WUMSTCDLAYQDNO-UHFFFAOYSA-N Synonym: hexyltriethoxysilane,triethoxy hexyl silane,hexyl triethoxysilane,n-hexyltriethoxysilane,hexyl-triethoxysilane,acmc-209ei3 PubChem CID: 4437318 IUPAC Name: triethoxy(hexyl)silane SMILES: CCCCCC[Si](OCC)(OCC)OCC