accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SiCH3ClN

CAS RN 18156-15-5

IUPAC Name: 4-(chlorodimethylsilyl)butanenitrile
Synonyms: 4-(chlorodimethylsilyl)butanenitrile
Molecular Weight (g/mol): 161.7
Molecular Formula: C6H12ClNSi
InChi Key: GPIARXZSVWTOMD-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CCCC#N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)