CAS RN 18096-70-3
IUPAC Name:
3-(dimethylamino)-1-(4-methoxyphenyl)prop-2-en-1-one
Synonyms:
3-(dimethylamino)-1-(4-methoxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol):
205.26
Molecular Formula:
C12H15NO2
InChi Key:
FXNAUCNJLHEGGF-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C(=O)C=CN(C)C
1
–
1
of
1
results
1
3-Dimethylamino-1-(4-methoxyphenyl)-2-propen-1-one, 95%, Thermo Scientific™
CAS: 18096-70-3 Molecular Formula: C12H15NO2 Molecular Weight (g/mol): 205.26 MDL Number: MFCD00097914,MFCD00097914 InChI Key: FXNAUCNJLHEGGF-UHFFFAOYSA-N Synonym: 2z-3-dimethylamino-1-4-methoxyphenyl prop-2-en-1-one,3-dimethylamino-1-4-methoxy-phenyl-propenone PubChem CID: 17024616 IUPAC Name: (Z)-3-(dimethylamino)-1-(4-methoxyphenyl)prop-2-en-1-one SMILES: COC1=CC=C(C=C1)C(=O)C=CN(C)C