accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 18039-90-2

OOOClTiClCl

CAS RN 18039-90-2

IUPAC Name: tris(oxolane); trichlorotitanium
Synonyms: tris(THF); titanium(III) chloride
Molecular Weight (g/mol): 370.54
Molecular Formula: C12H24Cl3O3Ti
InChi Key: MIDYWIPTCYRMQF-UHFFFAOYSA-K
SMILES: Cl[Ti](Cl)Cl.C1CCOC1.C1CCOC1.C1CCOC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results