accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 18039-26-4

IUPAC Name: 3,4,5-tris(benzyloxy)oxan-2-ol
Synonyms: 3,4,5-tris(benzyloxy)oxan-2-ol
Molecular Weight (g/mol): 420.51
Molecular Formula: C26H28O5
InChi Key: HTSKDJMXBBFKKG-UHFFFAOYNA-N
SMILES: OC1OCC(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results