accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONHNH2Br

CAS RN 180340-70-9

IUPAC Name: 2-amino-3-bromo-5-(methoxycarbonyl)pyridin-1-ium
Synonyms: 2-amino-3-bromo-5-(methoxycarbonyl)pyridin-1-ium
Molecular Weight (g/mol): 232.06
Molecular Formula: C7H8BrN2O2
InChi Key: LNTAAMVZKGYIIA-UHFFFAOYSA-O
SMILES: COC(=O)C1=C[NH+]=C(N)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results