accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNNNCH3

CAS RN 179873-47-3

IUPAC Name: methyl({[4-(1-methyl-1H-pyrazol-3-yl)phenyl]methyl})amine
Synonyms: methyl({[4-(1-methylpyrazol-3-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 201.27
Molecular Formula: C12H15N3
InChi Key: BBOCGPKUAXCMLG-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)C1=NN(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results