accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OSiH3CCH3OH3COH3C

CAS RN 17980-47-1

IUPAC Name: triethoxy(2-methylpropyl)silane
Synonyms: triethoxy(2-methylpropyl)silane
Molecular Weight (g/mol): 220.38
Molecular Formula: C10H24O3Si
InChi Key: ALVYUZIFSCKIFP-UHFFFAOYSA-N
SMILES: CCO[Si](CC(C)C)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results