CAS RN 1794783-36-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac 3,4-Dihydroxyphenylethylene Glycol-d5, TRC
CAS: 1794783-36-0 Molecular Formula: C8H5D5O4 Molecular Weight (g/mol): 175.19 Synonym: DL-3,4-Dihydroxyphenylglycol-d5,(+/-)-4-(1,2-Dihydroxyethyl)pyrocatechol-d5,4-(1,2-Dihydroxyethyl)-1,2-benzenediol-d5 SMILES: OC1=C(O)C([2H])=C(C(O)C([2H])([2H])O)C([2H])=C1[2H]