CAS RN 1788033-05-5
CAS RN 1788033-05-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1S,2S,3R,5S)-3-[[3-[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]-5-(propylthio)-3H-1,2,3-triazolo[4,5-d]pyrimidin-7-yl]amino]-5-(2-hydroxyethoxy)-1,2-cyclopentanediol, TRC
CAS: 1788033-05-5 Molecular Formula: C23H28F2N6O4S Molecular Weight (g/mol): 522.57 Synonym: Ticagrelor Imp A SMILES: CCCSc1nc(N[C@@H]2C[C@H](OCCO)[C@@H](O)[C@H]2O)c3nnn([C@@H]4C[C@H]4c5ccc(F)c(F)c5)c3n1