accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SOOHNNH2

CAS RN 17852-67-4

IUPAC Name: (4-methanesulfonylphenyl)hydrazine
Synonyms: (4-methanesulfonylphenyl)hydrazine
Molecular Weight (g/mol): 186.23
Molecular Formula: C7H10N2O2S
InChi Key: ZHYAENSFCNMJQQ-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(NN)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results