CAS RN 1781848-13-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Chloro-6-fluoro-2-methoxybenzoic Acid, TRC
CAS: 1781848-13-2 Molecular Formula: C8H6O3FCl Molecular Weight (g/mol): 204.58 IUPAC Name: 3-chloro-6-fluoro-2-methoxybenzoic acid SMILES: COc1c(Cl)ccc(F)c1C(=O)O