CAS RN 178176-80-2
IUPAC Name:
2-(diphenylphosphanyl)naphthalene-1-carboxylic acid
Synonyms:
2-(diphenylphosphanyl)naphthalene-1-carboxylic acid
Molecular Weight (g/mol):
356.36
Molecular Formula:
C23H17O2P
InChi Key:
WJCBDBBZIGUHOB-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Diphenylphosphino-1-naphthoic acid, 97%, Thermo Scientific Chemicals
CAS: 178176-80-2 Molecular Formula: C23H17O2P Molecular Weight (g/mol): 356.36 MDL Number: MFCD02683090 InChI Key: WJCBDBBZIGUHOB-UHFFFAOYSA-N Synonym: 2-diphenylphosphino-1-naphthoic acid,2-diphenylphosphanyl naphthalene-1-carboxylic acid,acmc-20anx1,2-diphenylphosphino-1-naphthalenecarboxylic acid,2-diphenylphosphino-naphthalen-1-carboxylic acid,1-naphthalenecarboxylicacid, 2-diphenylphosphino PubChem CID: 10861198 IUPAC Name: 2-diphenylphosphanylnaphthalene-1-carboxylic acid SMILES: OC(=O)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1