accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 177995-40-3

IUPAC Name: 4-methoxy-1,2-benzoxazol-3-amine
Synonyms: 4-methoxy-1,2-benzoxazol-3-amine
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: YFNIGSJHKGKXEQ-UHFFFAOYSA-N
SMILES: COC1=C2C(N)=NOC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results