CAS RN 1778-09-2
CAS RN 1778-09-2
IUPAC Name:
1-[4-(methylsulfanyl)phenyl]ethan-1-one
Synonyms:
1-[4-(methylsulfanyl)phenyl]ethanone
Molecular Weight (g/mol):
166.24
Molecular Formula:
C9H10OS
InChi Key:
JECUZQLBQKNEMW-UHFFFAOYSA-N
SMILES:
CSC1=CC=C(C=C1)C(C)=O
1
–
2
of
2
results
1
4'-(Methylthio)acetophenone, 99%
CAS: 1778-09-2 Molecular Formula: C9H10OS Molecular Weight (g/mol): 166.24 MDL Number: MFCD00039835 InChI Key: JECUZQLBQKNEMW-UHFFFAOYSA-N Synonym: 4'-methylthio acetophenone,4-methylthio acetophenone,4'-methylthioacetophenone,1-4-methylsulfanyl phenyl ethanone,4-acetyl thioanisole,4'-methylmercapto acetophenone,1-4-methylthio phenyl ethanone,1-4-methylthio phenyl ethan-1-one,1-4-methylsulfanylphenyl ethanone PubChem CID: 74501 IUPAC Name: 1-(4-methylsulfanylphenyl)ethanone SMILES: CSC1=CC=C(C=C1)C(C)=O