accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17773-10-3

ICH3NCH3CH3HO

CAS RN 17773-10-3

IUPAC Name: (2-hydroxyethyl)trimethylazanium iodide
Synonyms: choline iodide
Molecular Weight (g/mol): 231.08
Molecular Formula: C5H14INO
InChi Key: FNPBHXSBDADRBT-UHFFFAOYSA-M
SMILES: [I-].C[N+](C)(C)CCO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results