CAS RN 177662-76-9
IUPAC Name:
4-methanesulfonylaniline
Synonyms:
4-methylsulfonylaniline
Molecular Weight (g/mol):
171.21
Molecular Formula:
C7H9NO2S
InChi Key:
XJEVFFNOMKXBLU-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)C1=CC=C(N)C=C1
1
–
2
of
2
results
2
4-(Methylsulfonyl)aniline hydrochloride, 99%
CAS: 177662-76-9 Molecular Formula: C7H9NO2S Molecular Weight (g/mol): 171.21 MDL Number: MFCD00216490 InChI Key: XJEVFFNOMKXBLU-UHFFFAOYSA-N Synonym: 4-methylsulfonylaniline hydrochloride,4-methylsulfonyl aniline hydrochloride,4-methanesulfonylaniline hydrochloride,4-methylsulphonylaniline hydrochloride,4-methylsulphonyl aniline hydrochloride,4-methanesulfonyl aniline hydrochloride,4 methanesulfonyl aniline hydrochloride,acmc-1c3bo,4-methanesulonyl aniline hydrochloride,4-methanesulfonylphenylamine hydrochloride PubChem CID: 2735180 IUPAC Name: 4-methanesulfonylaniline SMILES: CS(=O)(=O)C1=CC=C(N)C=C1