accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 177172-49-5

ClH3CO(R)CH3NHClH

CAS RN 177172-49-5

IUPAC Name: hydrogen [3-(2-chlorophenyl)propyl][(1R)-1-(3-methoxyphenyl)ethyl]amine chloride
Synonyms: hydrogen [3-(2-chlorophenyl)propyl][(1R)-1-(3-methoxyphenyl)ethyl]amine chloride
Molecular Weight (g/mol): 340.29
Molecular Formula: C18H23Cl2NO
InChi Key: YJXUXANREVNZLH-PFEQFJNWSA-N
SMILES: [H+].[Cl-].COC1=CC=CC(=C1)[C@@H](C)NCCCC1=CC=CC=C1Cl
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results