accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17682-70-1

IUPAC Name: {4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[6.4.0.0²,⁶]dodecan-6-yl}methanol
Synonyms: {4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[6.4.0.0²,⁶]dodecan-6-yl}methanol
Molecular Weight (g/mol): 260.29
Molecular Formula: C12H20O6
InChi Key: GQXSDDHYUVYJCQ-UHFFFAOYNA-N
SMILES: CC1(C)OC2C3OC(C)(C)OCC3OC2(CO)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results