accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ON

CAS RN 176690-44-1

IUPAC Name: 2-(pyridin-3-yl)benzaldehyde
Synonyms: 2-(pyridin-3-yl)benzaldehyde
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: DTUANRRVVJRTJS-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1C1=CC=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results