accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17630-76-1

ClHNOO

CAS RN 17630-76-1

IUPAC Name: 5-chloro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-chloro-1H-indole-2,3-dione
Molecular Weight (g/mol): 181.58
Molecular Formula: C8H4ClNO2
InChi Key: XHDJYQWGFIBCEP-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC(=O)C(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results