accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1762-34-1

CH3NCH3N

CAS RN 1762-34-1

IUPAC Name: 5,5'-dimethyl-2,2'-bipyridine
Synonyms: 5,5'-dimethyl-2,2'-bipyridine
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: PTRATZCAGVBFIQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(N=C1)C1=CC=C(C)C=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results