CAS RN 176032-78-3
CAS RN 176032-78-3
IUPAC Name:
3-(1H-1,2,4-triazol-1-yl)aniline
Synonyms:
3-(1,2,4-triazol-1-yl)aniline
Molecular Weight (g/mol):
160.18
Molecular Formula:
C8H8N4
InChi Key:
QOHJDIAWBVPZFL-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(=C1)N1C=NC=N1
1
–
1
of
1
results
1
3-(1h-1,2,4-Triazol-1-yl)aniline, 90%, Thermo Scientific™
CAS: 176032-78-3 Molecular Formula: C8H8N4 Molecular Weight (g/mol): 160.18 InChI Key: QOHJDIAWBVPZFL-UHFFFAOYSA-N PubChem CID: 23503656 IUPAC Name: 3-(1,2,4-triazol-1-yl)aniline SMILES: C1=CC(=CC(=C1)N)N2C=NC=N2