accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOHNOONaNa

CAS RN 17593-73-6

IUPAC Name: 2-[(carboxylatomethyl)azaniumyl]acetate
Synonyms: ammoniodiacetate
Molecular Weight (g/mol): 132.1
Molecular Formula: C4H6NO4
InChi Key: NBZBKCUXIYYUSX-UHFFFAOYSA-M
SMILES: [O-]C(=O)C[NH2+]CC([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results