accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 175883-62-2

H3COH3CBOHHO

CAS RN 175883-62-2

IUPAC Name: (4-methoxy-3-methylphenyl)boronic acid
Synonyms: 4-methoxy-3-methylphenylboronic acid
Molecular Weight (g/mol): 165.98
Molecular Formula: C8H11BO3
InChi Key: PXVDQGVAZBTFIB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C)B(O)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)