accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH3OHO

CAS RN 1758-25-4

IUPAC Name: 2-(2,5-dimethoxyphenyl)acetic acid
Synonyms: (2,5-dimethoxyphenyl)acetic acid
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: BBZDYQUXRFATHZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(CC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results