accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HBrBrON

CAS RN 17570-98-8

IUPAC Name: hydrogen 2-bromo-1-(pyridin-2-yl)ethan-1-one bromide
Synonyms: hydrogen 2-bromo-1-(pyridin-2-yl)ethanone bromide
Molecular Weight (g/mol): 280.95
Molecular Formula: C7H7Br2NO
InChi Key: BYKVUGZUYJUSKD-UHFFFAOYSA-N
SMILES: [H+].[Br-].BrCC(=O)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results