accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17547-79-4

IUPAC Name: 2-(4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid
Synonyms: (4-oxo-3,5-dihydro-2H-1,5-benzothiazepin-3-yl)acetic acid
Molecular Weight (g/mol): 237.27
Molecular Formula: C11H11NO3S
InChi Key: VCSCQLDBPVHQPW-UHFFFAOYNA-N
SMILES: OC(=O)CC1CSC2=CC=CC=C2NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results