accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OO(S)NNHOO

CAS RN 175453-07-3

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(pyridin-3-yl)propanoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(pyridin-3-yl)propanoate
Molecular Weight (g/mol): 387.42
Molecular Formula: C23H19N2O4
InChi Key: JQLPMTXRCLXOJO-NRFANRHFSA-M
SMILES: [O-]C(=O)[C@H](CC1=CC=CN=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results