accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COPOOCH3

CAS RN 1754-47-8

IUPAC Name: dioctyl phenylphosphonate
Synonyms: dioctyl phenylphosphonate
Molecular Weight (g/mol): 382.53
Molecular Formula: C22H39O3P
InChi Key: HAKMAMKAFTZXOZ-UHFFFAOYSA-N
SMILES: CCCCCCCCOP(=O)(OCCCCCCCC)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results