accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OSOBr

CAS RN 175278-64-5

IUPAC Name: 4-bromobenzene-1-sulfinate
Synonyms: 4-bromobenzenesulfinate
Molecular Weight (g/mol): 220.06
Molecular Formula: C6H4BrO2S
InChi Key: QBLQHDHWTVMHRH-UHFFFAOYSA-M
SMILES: [O-]S(=O)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results