accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CIH3C

CAS RN 175277-95-9

IUPAC Name: 1-ethyl-2-iodo-3-methylbenzene
Synonyms: 1-ethyl-2-iodo-3-methylbenzene
Molecular Weight (g/mol): 246.09
Molecular Formula: C9H11I
InChi Key: PCSPENCTMKMGFE-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(C)=C1I
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)