accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 175205-91-1

CH3HNH3COOCH3O

CAS RN 175205-91-1

IUPAC Name: methyl 4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Synonyms: methyl 4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: OHVCRNQTGBXAPW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)NC(C)=C1C=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)