accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 175205-53-5

IUPAC Name: 4-(1,2,3-thiadiazol-4-yl)benzamide
Synonyms: 4-(1,2,3-thiadiazol-4-yl)benzamide
Molecular Weight (g/mol): 205.24
Molecular Formula: C9H7N3OS
InChi Key: PAVNUEROKSNSDS-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1)C1=CSN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results