accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NClClH3C

CAS RN 175205-50-2

IUPAC Name: 1-(3,5-dichlorophenyl)-2,5-dimethyl-1H-pyrrole
Synonyms: 1-(3,5-dichlorophenyl)-2,5-dimethylpyrrole
Molecular Weight (g/mol): 240.13
Molecular Formula: C12H11Cl2N
InChi Key: PZQGRGDARNXLSP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N1C1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results